Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium thiocyanate, 99+%, extra pure
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic Drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 149.0°C |
Color | White |
pH | 4.0 to 6.0 (5% aq. soln.) |
Physical Form | Crystals |
Quantity | 500 g |
Assay Percent Range | 99% min. (on dried basis) (Argentometry) |
Linear Formula | NH4SCN |
Merck Index | 15,558 |
Solubility Information | Solubility in water: 1600g/L (20°C). Other solubilities: soluble in alcohol, acetone and ammonia, practically insoluble in chcl3 and ethyl acetate |
Formula Weight | 76.12 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes serious eye damage. Harmful to aquatic life with long lasting effects. Contact with acids liberates very toxic gas. |
Precautionary Statement | GHS P Statement Keep container tightly closed. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Wash with plenty of soap and water. IF |
EINECSNumber | 217-175-6 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction