Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium thiocyanate, ACS, 97.5% min, Alfa Aesar™
Click to view available options
Quantity:
250g
1kg
Specifications
Assay Percent Range | 97.5% min |
Beilstein | 3595135 |
Solubility Information | Soluble in water,ethanol,methanol,ammonia,and acetone. Practically insoluble in CHCl3and ethyl acetate. |
Formula Weight | 76.12 |
Physical Form | Crystalline |
Grade | ACS |
Density | 1.305 |
Sensitivity | Hygroscopic; Light sensitive |
Melting Point | 147°C to 149°C |
Quantity | 250g |
Show More |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P363-P501c
H302+H312+H332-H318
EINECSNumber : 217-175-6
RTECSNumber : XK7875000
TSCA : Yes
Recommended Storage : Ambient temperatures