Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AMN 082 dihydrochloride
GREENER_CHOICE

Catalog No. 238510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-851-0 10 mg
23-855-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-851-0 Supplier Tocris Bioscience™ Supplier No. 2385/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective mGlu7 agonist

AMN 082 dihydrochloride is a the first selective mGlu7 agonist. Potently inhibits cAMP accumulation and stimulates GTPγS binding in recombinant cells and on membranes expressing mGlu7 (EC50 = 64 - 290 nM). Selective over other mGluR subtypes and selected ionotropic glutamate receptors up to 10 μM. Acts via a novel allosteric site and is orally active and brain penetrant. Reduces haloperidol-induced catalepsy in rats.

View important information regarding the usage of AMN 082 dihydrochloride .

Chemical Identifiers

CAS 97075-46-2
Molecular Formula C28H30Cl2N2

Specifications

Quantity 10 mg
Formula Weight 465.45
Purity >99%
Molecular Weight (g/mol) 465.462
Chemical Name or Material AMN 082 dihydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.