Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amodiaquine Antibody (6D10) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP105106 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105106 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105106 Supplier Novus Biologicals Supplier No. NBP105106

Mouse Monoclonal Antibody

Amodiaquine Monoclonal specifically detects Amodiaquine in Non-species specific samples. It is validated for ELISA.

Specifications

Antigen Amodiaquine
Applications ELISA
Classification Monoclonal
Clone 6D10
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:4000
Host Species Mouse
Immunogen 4-amino-2(diethylaminomethyl)phenol a preparation of a carrier protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for the non-quinoline sidechain of amodiaquine. Can be used for quantitation of amodiaquine in human serum or plasma with a detection limit of app. 2 ng/ml.
Target Species Non-species specific
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.