Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMOT Antibody, Novus Biologicals™
SDP

Catalog No. NBP178209 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178209 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178209 Supplier Novus Biologicals Supplier No. NBP178209

Rabbit Polyclonal Antibody

AMOT Polyclonal specifically detects AMOT in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen AMOT
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 mcg/mg
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_001106962.1
Gene Alias angiomotin, angiomotin p80 isoform, KIAA1071angiomotin p130 isoform
Gene Symbols AMOT
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 750 and 800 of human Angiomotin using the numbering given in entry NP_001106962.1 (GeneID 154796).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154796
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.