Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ampco Safety Tools Fireman's Hooks

$932.00 - $1018.00
Specifications
| Product Type | Fireman's Hook/Pike Pole |
|---|---|
| Material | Aluminum Bronze |
| For Use With (Application) | Hazardous, flammable or combustible environment |
| Catalog Number | Mfr. No. | Length (English) | Length (Metric) | Weight (English) | Weight (Metric) | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Length (English) | Length (Metric) | Weight (English) | Weight (Metric) | Price | Quantity & Availability | ||
19-166-444
![]() |
Ampco Safety Tools
PP48D |
48 in. | 121.92 cm | 7.6 lb. | 3.44 kg |
Each for $1,018.00
|
|
||
19-166-446
![]() |
Ampco Safety Tools
PP96 |
96 in. | 243.84 cm | 7.6 lb. | 3.44 kg |
Each for $992.50
|
|
||
19-166-445
![]() |
Ampco Safety Tools
PP72 |
72 in. | 182.88 cm | 6.5 lb. | 3 kg |
Each for $932.00
|
|
||
Specifications
| Fireman's Hook/Pike Pole | |
| Aluminum Bronze | |
| Hazardous, flammable or combustible environment |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title