Description
Includes
Four-piece tool kit
(19-805-933) includes: standard screwdriver (5/16 in. tip, 3 in. blade), 8 in. adjustable end wrench, 9 in. crate opener, 6.50 in. combination pliers.
11-piece tool kit
(19-805-934) includes: two standard screwdrivers (5/16 in. tip, 3 in. blade and5/16 in. tip, 6 in. blade), three adjustable end wrenches (6, 8, and 10 in.), 9 in. crate opener, No. 10 groove joint pliers, two No. 2 Phillips screwdriver, 10 in. adjustable pipe wrench, 16 oz. claw hammer with fiberglass handle.
16-piece tool kit
(19-120-3787) includes: four double end box wrenches (5/16 x 3/8 in., 7/16 x 1/2 in., 9/16 x 5/8 in., 3/4 x 7/8 in.), No. 2 Phillips screwdriver, standard screwdriver (5/16 in. tip, 6 in. blade), 10 in. adjustable wrench, two combination pliers (61/2 and 8 in.), 10 in. groove joint pliers, common knife (31/8 in. blade, 63/4 in. OAL), stiff putty knife (11/4 in.
- Specially engineered for use in safety-sensitive applications to reduce the risk of fire and explosions
- Nonsparking, nonmagnetic, and corrosion resistant
- Made in the USA
Certifications
FM approved.
Compliance
Designed using Federal, military, ANSI™, and DIN standards.
Specifications
Specifications
| Product Type | 17-piece Hazmat Tool Kit |
| For Use With (Application) | Safety-sensitive applications to reduce the risk of fire and explosions |
| Includes | B-399 Shoe handle brush, CJ-1-ST 9 in. crate opener, H-20FG 16 oz. claw hammer with Fiberglass handle, K-5 common knife, 5.25 in. blade, 10.25 in. OAL, K-21 putty knife, 1.56 x 3.56 in. blade, 7.75 in. OAL, P-312 12 in. groove joint plier, P-326 7 in. needle nose pliers with side cutter, P-31 8 in. adjustable combination pliers, S-11G deck scraper, 1.5 in. blade x 15 in. L, S-40 spray booth scraper, 3 in. blade x 9.25 in. long, S-49 standard screwdriver, 0.31 in. tip, 6 in. blade, S-1100 No. 3 Phillips screwdriver, 6 in. blade, S-1144 12 in. tin snip, W-71 8 in. adjustable end wrench, W-73 12in. adjustable end wrench, W-212AL 14 in. adjustable pipe wrench, W-56 12 in. bung wrench, Tool Bucket with Divider |
| Weight (English) | 19.9 lb. |
| Weight (Metric) | 9 kg |