Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPD1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224509SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224509SS 0.025 mg
NBP224509 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224509SS Supplier Novus Biologicals Supplier No. NBP224509SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

AMPD1 Polyclonal specifically detects AMPD1 in Bovine, Canine, Human, Mouse, Opossum, Primate, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen AMPD1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias adenosine monophosphate deaminase 1, adenosine monophosphate deaminase 1 (isoform M), adenosine monophosphate deaminase-1 (muscle), AMP deaminase 1, AMP deaminase isoform M, AMPD, EC 3.5.4.6, MAD, MADA, Myoadenylate deaminase, skeletal muscle AMPD
Gene Symbols AMPD1
Host Species Rabbit
Immunogen A portion of amino acids 140-190 of human AMPDA1 was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 270
Test Specificity 100% homologous in human (isoforms CRA_a and CRA_c), mouse, rat (isoforms CRA_a and CRA_b).
Target Species Bovine, Canine, Human, Mouse, Opossum, Primate, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.