Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amphiregulin Antibody (RM0151-7D17), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB361443 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361443 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361443 Supplier Novus Biologicals Supplier No. NBP212036APC

Rat Monoclonal Antibody

Amphiregulin Monoclonal antibody specifically detects Amphiregulin in Mouse samples. It is validated for Western Blot

Specifications

Antigen Amphiregulin
Applications Western Blot
Classification Monoclonal
Clone RM0151-7D17
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias amphiregulin, AR, AREG, AREGB, Colorectum cell-derived growth factor, CRDGF, MGC13647, schwannoma-derived growth factor, SDGF
Host Species Rat
Immunogen Mouse recombinant Amphiregulin
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Biology, Cytokine Research, Growth and Development, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 374
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.