Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amphiregulin Antibody (RM0151-7D17) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB019008 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019008 0.025 mg
NBP212036 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019008 Supplier Novus Biologicals Supplier No. NBP2120360.025MG

Rat Monoclonal Antibody

Amphiregulin Monoclonal specifically detects Amphiregulin in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Amphiregulin
Applications Western Blot
Classification Monoclonal
Clone RM0151-7D17
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. P31955
Gene Alias amphiregulin, AR, AREG, AREGB, Colorectum cell-derived growth factor, CRDGF, MGC13647, schwannoma-derived growth factor, SDGF
Gene Symbols AREG
Host Species Rat
Immunogen Mouse Amphiregulin
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cytokine Research, Growth and Development, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 374
Test Specificity This antibody detects mouse Amphiregulin in Western blotting.
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.