Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ampicillin, Anhydrous, 98.24%, MP Biomedicals™

Catalog No. ICN19014625 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
5 g
25 g
100 g

Catalog No. ICN19014625

Supplier: MP Biomedicals, Inc 0219014625

Chemical Identifiers

CAS 69-53-4
Molecular Formula C16H19N3O4S
Molecular Weight (g/mol) 349.405
InChI Key AVKUERGKIZMTKX-NJBDSQKTSA-N
Synonym Aminobenzylpenicillin
PubChem CID 6249
ChEBI CHEBI:28971
IUPAC Name (2S,5R,6R)-6-[[(2R)-2-amino-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMILES CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)N)C(=O)O)C

Specifications

Type Ampicillin, Anhydrous
Physical Form Powder
Color White
Quantity 25 g
Assay Percent Range 0.9824
Formula Weight 371.4
Chemical Name or Material Ampicillin, Anhydrous, For HPLC analysis
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.