Learn More
Description
Specifications
Specifications
| Antigen | AMPK alpha 1/2 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2500-1:5000, ELISA 1:50000, Immunohistochemistry 1:50-1:250, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:50-1:250 |
| Formulation | Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide |
| Gene Alias | AMPK, AMPKa1, PRKAA1 protein kinase, AMP-activated, alpha 1 catalytic subunit |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide taken within amino acid region 480-530 on rat 5'-AMP-activated protein kinase Felinealytic subunit alpha-2 protein. |
| Purification Method | Affinity-purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
