Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMPK alpha 1/2, p Thr172 Antibody (SR2077), Novus Biologicals™

Rabbit Monoclonal Antibody
$334.00 - $448.78
Specifications
| Antigen | AMPK alpha 1/2 |
|---|---|
| Clone | SR2077 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
AMPK alpha 1/2 Monoclonal antibody specifically detects AMPK alpha 1/2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| AMPK alpha 1/2 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AMPK, AMPKa1, PRKAA1 protein kinase, AMP-activated, alpha 1 catalytic subunit | |
| A synthesized peptide derived from human phospho-AMPK alpha 1/2 [Thr172] (Uniprot #: Q13131, P54646) | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| SR2077 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 5562 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title