Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK alpha 1/2, p Thr172 Antibody (SR2077), Novus Biologicals™
SDP

Catalog No. NB286792 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB286792 50 μg
NB283063 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB286792 Supplier Novus Biologicals Supplier No. NBP32153750UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

AMPK alpha 1/2 Monoclonal antibody specifically detects AMPK alpha 1/2 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen AMPK alpha 1/2
Applications Western Blot
Classification Monoclonal
Clone SR2077
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias AMPK, AMPKa1, PRKAA1 protein kinase, AMP-activated, alpha 1 catalytic subunit
Host Species Rabbit
Immunogen A synthesized peptide derived from human phospho-AMPK alpha 1/2 [Thr172] (Uniprot #: Q13131, P54646)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 5562
Target Species Human, Mouse, Rat
Content And Storage Store at -20 degrees C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.