Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK alpha 1 Antibody (2B7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP222127SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222127SS 0.025 mL
NBP222127 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP222127SS Supplier Novus Biologicals Supplier No. NBP222127SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 9 publications

AMPK alpha 1 Monoclonal specifically detects AMPK alpha 1 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen AMPK alpha 1
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2B7
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 μg/mL, Simple Western 1:50, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 2 - 4 μg/mL, Immunohistochemistry-Paraffin 1:200-1:1000
Gene Accession No. Q13131
Gene Alias AMP -activate kinase alpha 1 subunit, AMP-activated protein kinase, catalytic, alpha-1,5'-AMP-activated protein kinase catalytic subunit alpha-1, AMPK, AMPK alpha 1,5'-AMP-activated protein kinase, catalytic alpha-1 chain, AMPK subunit alpha-1, AMPK1, AMPKa1, EC 2.7.11, EC 2.7.11.1, MGC33776, MGC57364, protein kinase, AMP-activated, alpha 1 catalytic subunit
Gene Symbols PRKAA1
Host Species Mouse
Immunogen Purified recombinant fragment of human AMPK alpha 1 expressed in E. coli. [UniProt# Q13131]
Molecular Weight of Antigen 64 kDa
Purification Method Protein A or G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Signal Transduction, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 5562
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Precipitation
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.