Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK alpha 1 Antibody (2B7), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N222127PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N222127PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N222127PEC7 Supplier Novus Biologicals Supplier No. NBP222127PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AMPK alpha 1 Monoclonal specifically detects AMPK alpha 1 in Human, Mouse, Rat, Primate samples. It is validated for Flow Cytometry.

Specifications

Antigen AMPK alpha 1
Applications Flow Cytometry
Classification Monoclonal
Clone 2B7
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias AMP -activate kinase alpha 1 subunit, AMP-activated protein kinase, catalytic, alpha-1,5′-AMP-activated protein kinase catalytic subunit alpha-1, AMPK, AMPK alpha 1,5′-AMP-activated protein kinase, catalytic alpha-1 chain, AMPK subunit alpha-1, AMPK1, AMPKa1, EC 2.7.11, EC 2.7.11.1, MGC33776, MGC57364, protein kinase, AMP-activated, alpha 1 catalytic subunit
Gene Symbols PRKAA1
Host Species Mouse
Immunogen Purified recombinant fragment of human AMPK alpha 1 expressed in E. coli. [UniProt No. Q13131]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Research Discipline Apoptosis, Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Signal Transduction, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 5562.0
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.