Learn More
Description
Specifications
Specifications
| Antigen | AMPK alpha 1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 750 |
| Dilution | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | AMP -activate kinase alpha 1 subunit, AMP-activated protein kinase, catalytic, alpha-1,5′-AMP-activated protein kinase catalytic subunit alpha-1, AMPK, AMPK alpha 1,5′-AMP-activated protein kinase, catalytic alpha-1 chain, AMPK subunit alpha-1, AMPK1, AMPKa1, EC 2.7.11, EC 2.7.11.1, MGC33776, MGC57364, protein kinase, AMP-activated, alpha 1 catalytic subunit |
| Gene Symbols | PRKAA1 |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residues 350 and 400 of human 5′-AMP-activated Protein Kinase, catalytic Alpha-1 chain using the numbering given in entry BAA36547.1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
