Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMPK alpha 1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP315610100UL
This item is not returnable.
View return policy
Description
AMPK alpha 1 Polyclonal antibody specifically detects AMPK alpha 1 in Human samples. It is validated for Western Blot, KnockDownSpecifications
AMPK alpha 1 | |
Polyclonal | |
Western Blot 1:500 - 1:2000, Knockout Validated | |
AMP -activate kinase alpha 1 subunit, AMP-activated protein kinase, catalytic, alpha-1,5'-AMP-activated protein kinase catalytic subunit alpha-1, AMPK, AMPK alpha 1,5'-AMP-activated protein kinase, catalytic alpha-1 chain, AMPK subunit alpha-1, AMPK1, AMPKa1, EC 2.7.11, EC 2.7.11.1, MGC33776, MGC57364, protein kinase, AMP-activated, alpha 1 catalytic subunit | |
Recombinant protein of human AMPK alpha 1. | |
100 μg | |
Apoptosis, Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Signal Transduction, Translation Control | |
5562 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, KnockDown | |
Unconjugated | |
PBS, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction