Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AMPK alpha 1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$171.20 - $458.00
Specifications
Antigen | AMPK alpha 1 |
---|---|
Dilution | Western Blot 1:500 - 1:2000, Knockout Validated |
Applications | Western Blot, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
AMPK alpha 1 Polyclonal antibody specifically detects AMPK alpha 1 in Human samples. It is validated for Western Blot, KnockDownSpecifications
AMPK alpha 1 | |
Western Blot, KnockDown | |
Unconjugated | |
Rabbit | |
Apoptosis, Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Signal Transduction, Translation Control | |
PBS, 50% glycerol, pH7.3 | |
5562 | |
IgG | |
Affinity purified |
Western Blot 1:500 - 1:2000, Knockout Validated | |
Polyclonal | |
Purified | |
RUO | |
Human | |
AMP -activate kinase alpha 1 subunit, AMP-activated protein kinase, catalytic, alpha-1,5'-AMP-activated protein kinase catalytic subunit alpha-1, AMPK, AMPK alpha 1,5'-AMP-activated protein kinase, catalytic alpha-1 chain, AMPK subunit alpha-1, AMPK1, AMPKa1, EC 2.7.11, EC 2.7.11.1, MGC33776, MGC57364, protein kinase, AMP-activated, alpha 1 catalytic subunit | |
Recombinant protein of human AMPK alpha 1. | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title