Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK alpha 2 Antibody (8E11H5) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261852 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61852 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61852 Supplier Novus Biologicals Supplier No. NBP261852
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AMPK alpha 2 Monoclonal specifically detects AMPK alpha 2 in Human, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen AMPK alpha 2
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8E11H5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
Formulation PBS with 0.05% Sodium Azide
Gene Alias 5′-AMP-activated protein kinase catalytic subunit alpha-2, AMPK subunit alpha-2, AMPK2, AMPK5′-AMP-activated protein kinase, catalytic alpha-2 chain, AMPK-alpha-2 chain, EC 2.7.11, EC 2.7.11.1, PRKAA, protein kinase, AMP-activated, alpha 2 catalytic subunit
Gene Symbols PRKAA2
Host Species Mouse
Immunogen Purified recombinant fragment of human PRKAA2 (AA: 453-552) expressed in E. Coli.
Molecular Weight of Antigen 62.3 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 5563
Target Species Human, Monkey
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.