Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK alpha 2, p Thr172 Antibody (4F4), Novus Biologicals™
SDP

Catalog No. NB298176 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298176 50 μL
NB298177 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298176 Supplier Novus Biologicals Supplier No. NBP32640050UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

AMPK alpha 2 Monoclonal antibody specifically detects AMPK alpha 2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry

Specifications

Antigen AMPK alpha 2
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 4F4
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias 5'-AMP-activated protein kinase catalytic subunit alpha-2, AMPK subunit alpha-2, AMPK2, AMPK5'-AMP-activated protein kinase, catalytic alpha-2 chain, AMPK-alpha-2 chain, EC 2.7.11, EC 2.7.11.1, PRKAA, protein kinase, AMP-activated, alpha 2 catalytic subunit
Host Species Rabbit
Immunogen A synthesized peptide derived from Human AMPK alpha 2 [p Thr172] [UniProt P54646]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Hypoxia, MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 5563
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.