Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK beta 2/PRKAB2 Antibody (OTI4H4), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP271226A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP271226A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP271226A70 Supplier Novus Biologicals Supplier No. NBP271226AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AMPK beta 2/PRKAB2 Monoclonal specifically detects AMPK beta 2/PRKAB2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen AMPK beta 2/PRKAB2
Applications Western Blot
Classification Monoclonal
Clone OTI4H4
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 5′-AMP-activated protein kinase, beta-2 subunit, AMPK beta 2,5′-AMP-activated protein kinase subunit beta-2, AMPK beta-2 chain, AMPK subunit beta-2, MGC61468, protein kinase, AMP-activated, beta 2 non-catalytic subunit
Gene Symbols PRKAB2
Host Species Mouse
Immunogen Full length recombinant protein of human AMPK beta 2/PRKAB2 (NP_005390) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5565.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.