Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK beta 2/PRKAB2 Antibody (OTI4H4), PerCP, Novus Biologicals™
SDP

Catalog No. NB289008 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB289008 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB289008 Supplier Novus Biologicals Supplier No. NBP271226PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

AMPK beta 2/PRKAB2 Monoclonal antibody specifically detects AMPK beta 2/PRKAB2 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen AMPK beta 2/PRKAB2
Applications Western Blot
Classification Monoclonal
Clone OTI4H4
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias 5'-AMP-activated protein kinase, beta-2 subunit, AMPK beta 2,5'-AMP-activated protein kinase subunit beta-2, AMPK beta-2 chain, AMPK subunit beta-2, MGC61468, protein kinase, AMP-activated, beta 2 non-catalytic subunit
Host Species Mouse
Immunogen Full length human recombinant protein of human PRKAB2 (NP_005390) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5565
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.