Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK gamma 1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15066805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15066805 0.05 mg
NB018658 0.025 mg
NBP150668 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP15066805 Supplier Novus Biologicals Supplier No. NBP1506680.05MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AMPK gamma 1 Polyclonal specifically detects AMPK gamma 1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen AMPK gamma 1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS with 0.09% sodium azide
Gene Accession No. P54619
Gene Alias 5′-AMP-activated protein kinase subunit gamma-1, 5′-AMP-activated protein kinase, gamma-1 subunit, AMPK gamma1, AMPK gamma-1 chain, AMPK subunit gamma-1, AMPKg, MGC8666, protein kinase, AMP-activated, gamma 1 non-catalytic subunit
Gene Symbols PRKAG1
Host Species Rabbit
Immunogen Synthetic peptide selected from the center of human PRKAG1
Quantity 0.05 mg
Primary or Secondary Primary
Gene ID (Entrez) 5571
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.