Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMPK gamma 1 Rabbit anti-Human, Mouse, Rat, Clone: S08-7D5, Novus Biologicals™
SDP

Catalog No. NB188344 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188344 100 μg
NB188345 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB188344 Supplier Novus Biologicals Supplier No. NBP314994100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

AMPK gamma 1 Monoclonal antibody specifically detects AMPK gamma 1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen AMPK gamma 1
Applications Western Blot
Classification Monoclonal
Clone S08-7D5
Conjugate Unconjugated
Formulation 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide
Gene Alias 5'-AMP-activated protein kinase subunit gamma-1, 5'-AMP-activated protein kinase, gamma-1 subunit, AMPK gamma1, AMPK gamma-1 chain, AMPK subunit gamma-1, AMPKg, MGC8666, protein kinase, AMP-activated, gamma 1 non-catalytic subunit
Host Species Rabbit
Immunogen A synthetic peptide of human AMPK gamma 1 (Uniprot#: P54619)
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 5571
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.