Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amylin Antibody, Novus Biologicals™
SDP

Catalog No. NBP16921020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP16921020 20 μL
NBP169210 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP16921020 Supplier Novus Biologicals Supplier No. NBP16921020UL

Rabbit Polyclonal Antibody

Amylin Polyclonal specifically detects Amylin in Human samples. It is validated for Western Blot.

Specifications

Antigen Amylin
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.2-1 ug/ml
Formulation From PBS & 2% Sucrose. with No Preservative
Gene Accession No. P10997
Gene Alias AMYLIN, DAPamylin, Diabetes-associated peptide, IAP, Insulinoma amyloid peptide, islet amyloid polypeptide, Islet amyloid polypeptide (diabetes-associated peptide; amylin)
Gene Symbols IAPP
Host Species Rabbit
Immunogen Synthetic peptides corresponding to IAPP (islet amyloid polypeptide) The peptide sequence was selected from the N terminal of IAPP (NP_000406). Peptide sequence MGILKLQVFLIVLSVALNHLKATPIESHQVEKRKCNTATCATQRLANFLV.
Molecular Weight of Antigen 4 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3375
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.