Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amylin Antibody (R10/99), Novus Biologicals™
SDP

Catalog No. NB018251 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018251 0.25 mL
NB600713 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018251 Supplier Novus Biologicals Supplier No. NB6007130.25ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 7 publications

Amylin Monoclonal specifically detects Amylin in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Amylin
Applications Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence
Classification Monoclonal
Clone R10/99
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100
Formulation Tissue culture supernatant with 0.09% Sodium Azide
Gene Accession No. P10997
Gene Alias AMYLIN, DAPamylin, Diabetes-associated peptide, IAP, Insulinoma amyloid peptide, islet amyloid polypeptide, Islet amyloid polypeptide (diabetes-associated peptide; amylin)
Gene Symbols IAPP
Host Species Mouse
Immunogen Synthetic peptide: CATQRLANFLV, corresponding to internal sequence amino acids 40-50 of Human Amylin coupled to tuberculin.
Purification Method Tissue culture supernatant
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3375
Test Specificity The is of high affinity, and stains islet amyloid peptide in intra-cellular insulin granules, intra-cellular amyloid and extra-cellular islet amyloid.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Supernatant
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.