Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amylin Antibody (R10/99), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 7 publications
$206.50 - $591.65
Specifications
| Antigen | Amylin |
|---|---|
| Clone | R10/99 |
| Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence |
| Classification | Monoclonal |
Description
Amylin Monoclonal specifically detects Amylin in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| Amylin | |
| Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100 | |
| Monoclonal | |
| Supernatant | |
| RUO | |
| Human | |
| AMYLIN, DAPamylin, Diabetes-associated peptide, IAP, Insulinoma amyloid peptide, islet amyloid polypeptide, Islet amyloid polypeptide (diabetes-associated peptide; amylin) | |
| IAPP | |
| IgG1 | |
| Tissue culture supernatant | |
| The is of high affinity, and stains islet amyloid peptide in intra-cellular insulin granules, intra-cellular amyloid and extra-cellular islet amyloid. |
| R10/99 | |
| Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Apoptosis, Cancer | |
| P10997 | |
| 3375 | |
| Synthetic peptide: CATQRLANFLV, corresponding to internal sequence amino acids 40-50 of Human Amylin coupled to tuberculin. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title