Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Amyloid beta fibrils Rabbit anti-Human, Mouse, Clone: M98, MilliporeSigma™
Rabbit Monoclonal Antibody
$197.00 - $493.00
Specifications
| Antigen | Amyloid beta fibrils | 
|---|---|
| Clone | M98 | 
| Applications | Immunohistochemistry (Paraffin) | 
| Classification | Monoclonal | 
| Conjugate | Unconjugated | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
| 
                
                
                    
                        MABN640I25
                    
                    
                        
                    
                    
                        
                         | 
            
                
                    
                        MilliporeSigma
                         MABN640I25UG  | 
                
            
            
            
            
            
                
                    25 μg | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $197.00
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
| 
                
                
                    
                        MABN640I100
                    
                    
                        
                    
                    
                        
                         | 
            
                
                    
                        MilliporeSigma
                         MABN640I100UG  | 
                
            
            
            
            
            
                
                    100 μg | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $493.00
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
Description
Amyloid beta fibrils Monoclonal antibody specifically detects Amyloid beta fibrils in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin).Specifications
| Amyloid beta fibrils | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Fibrillar Abeta 1-42 peptide | |
| Protein A purified | 
| M98 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Purified rabbit recombinant monoclonal antibody in PBS with 0.05% sodium azide. | |
| Primary | |
| Stable for 1 year at 2-8°C from date of receipt. | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title