Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Anacardic acid
GREENER_CHOICE

Catalog No. p-4783744
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Noncompetitive PCAF/p300 inhibitor

Noncompetitive inhibitor of PCAF and p300 histone acetyltransferase (HAT) activity (IC50 values are ∽5.0 and ∽8.5 μM respectively). Inhibits NF-κB activation. Also reported to selectively activate Aurora kinase A-mediated phosphorylation of histone H3 and to inhibit protein SUMOylation. Exhibits antitumor, antimicrobial and antioxidant activity.

Chemical Identifiers

CAS 16611-84-0
Molecular Formula C22H36O3

Specifications

Quantity 10 mg
Formula Weight 348.52
Purity >97%
Molecular Weight (g/mol) 348.527
Chemical Name or Material Anacardic acid
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.