Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANAPC13 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB044577 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB044577 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB044577 Supplier Novus Biologicals Supplier No. NBP297529G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ANAPC13 Polyclonal specifically detects ANAPC13 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ANAPC13
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias anaphase promoting complex subunit 13, anaphase-promoting complex subunit 13, APC13SWM1, cyclosome subunit 13, Cylosome subunit 13, DKFZP566D193, homolog of yeast Swm1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ANAPC13 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25847
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.