Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Andes Virus Glycoprotein Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB199894 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB199894 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB199894 Supplier Novus Biologicals Supplier No. NBP241253R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Andes Virus Glycoprotein Polyclonal antibody specifically detects Andes Virus Glycoprotein in Virus samples. It is validated for ELISA

Specifications

Antigen Andes Virus Glycoprotein
Applications ELISA
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias G1 and G2 surface glycoprotein precursor
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid peptide near the amino terminus of the Andes virus glycoprotein. The immunogen is located within amino acids 70 - 120 of Andes Virus Glycoprotein. Amino Acid Squence: VEWRKKSDTTDTTNAASTT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 991233
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.