Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Andes Virus Nucleocapsid Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB199985 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB199985 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB199985 Supplier Novus Biologicals Supplier No. NBP241254AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Andes Virus Nucleocapsid Polyclonal antibody specifically detects Andes Virus Nucleocapsid in Virus samples. It is validated for ELISA

Specifications

Antigen Andes Virus Nucleocapsid
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias Nucleocapsid protein
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid peptide near the amino terminus of the Andes virus nuclecapsid. The immunogen is located within amino acids 130 - 180 of Andes Virus Nucleocapsid. Amino Acid Squence: TRGRQTVKDNKGTRIRFKD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites)
Primary or Secondary Primary
Gene ID (Entrez) 991232
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.