Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Androgen R/NR3C4 Antibody (2621E), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB295993 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295993 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295993 Supplier Novus Biologicals Supplier No. FAB58762U

Rabbit Monoclonal Antibody

Androgen R/NR3C4 Monoclonal antibody specifically detects Androgen R/NR3C4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen Androgen R/NR3C4
Applications Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 2621E
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy
Host Species Rabbit
Immunogen Synthetic Human Androgen R/NR3C4 peptide
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cellular Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 367
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.