Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Androgen R/NR3C4 Antibody (523339), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB377409 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377409 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377409 Supplier Novus Biologicals Supplier No. FAB5876E

Mouse Monoclonal Antibody

Androgen R/NR3C4 Monoclonal antibody specifically detects Androgen R/NR3C4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen Androgen R/NR3C4
Applications Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 523339
Conjugate DyLight 405
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy
Host Species Mouse
Immunogen E. coli-derived recombinant human Androgen R/NR3C4, Thr660-Gln919, Accession # P10275
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 367
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.