Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Androgen R/NR3C4 Antibody (AR441 + DHTR/882), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB007477 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB007477 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB007477 Supplier Novus Biologicals Supplier No. NBP247857AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Androgen R/NR3C4 Monoclonal specifically detects Androgen R/NR3C4 in Human, Mouse (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Androgen R/NR3C4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone AR441 + DHTR/882
Conjugate Alexa Fluor 750
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy
Gene Symbols AR
Host Species Mouse
Immunogen A synthetic peptide, aa 302-318 of human Androgen R/NR3C4 (Androgen R/NR3C4441); Recombinant human DHTR protein (DHTR/882) (Uniprot: P10275)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cellular Markers, Phospho Specific
Primary or Secondary Primary
Test Specificity Recognizes a protein of 110kDa, which is identified as androgen receptor (AR). It reacts with full length, and the newly described A form of the receptor. It does not cross react with estrogen, progesterone, or glucocorticoid receptors. The expression of AR is reportedly inversely correlated with histologic grade i.e. well differentiated prostate tumors show higher expression than the poorly differentiated tumors. In prostate cancer, AR has been proposed, as a marker of hormone-responsiveness and thus it may be useful in identifying patients likely to benefit from anti-androgen therapy. Anti-androgen receptor has been useful clinically in differentiating morpheaform basal cell carcinoma (mBCC) from desmoplastic trichoepithelioma (DTE) in the skin. This monoclonal antibody is superb for staining of formalin/paraffin tissues.
Target Species Human, Mouse (Negative)
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.