Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Androgen R/NR3C4, p Ser210 Antibody (156C135.2), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. N106603A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N106603A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N106603A30 Supplier Novus Biologicals Supplier No. NB10056603AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Androgen R/NR3C4 Monoclonal specifically detects Androgen R/NR3C4 in Canine, Human, Mouse, Primate, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Androgen R/NR3C4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 156C135.2
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias AIS, androgen receptor, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy
Gene Symbols AR
Host Species Mouse
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids 207-221 (GRAREAS*GAPTSSKD) of human androgen receptor, containing the serine 213 phosphorylation site: GenBank Accession No. A39248. Note: S* refers to phosphorylated serine in the peptide sequence.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Breast Cancer, Cancer, Cellular Markers, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 367.0
Test Specificity In IGF-1 stimulated LNCaP cells (passage number 38), a ∼110 kDa band was observed. Please see Lin et al. 2003 for additional details. The serine phosphorylation site recognized by this antibody has been alternatively referred to Ser213 (Lee and Chang, 2003) and Ser210 (Lin et al, 2003). Variations in denotation can arise from how the sequence is counted in various GenBank accession numbers. The site is denoted as Ser213 in GenBank Accession No. A39248, which was used to design the immunogen.
Target Species Canine, Human, Mouse, Primate, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.