Learn More
Azure Biosystems Anesthesia Nose Cone
SDP
Catalog No. IS4050AZ Shop All Azure Biosystems Products
Change view
Click to view available options
For Use With (Equipment):
Sapphire FL
Catalog No. For Use With (Equipment)
IS4050AZ Sapphire FL
1 options
Catalog No. IS4050AZ Supplier Azure Biosystems Supplier No. IS4050
Only null left
Add to Cart
Add to Cart
Set of five 7 mm anesthesia nose cones for a tight fit with laboratory mice. For use with Anesthesia Tubing (IS4051).
  • Set of five 7 mm anesthesia nose cones for a tight fit with laboratory mice.
  • For use with Anesthesia Tubing (IS4051).
Specifications
For Use With (Equipment) Sapphire FL
Product Type Nose Cone