Learn More
Manganese(II) sulfate hydrate, Puratronic™, 99.999% (metals basis)
Description
Used in feed additive, nutrient, firming agent and flavor enhancer. It is also used as a fermentation aid in the processing of beer and malt beverages.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Pink |
| Physical Form | Crystalline Powder |
| Quantity | 25 g |
| Assay Percent Range | 99.999% (Metal basis) |
| Solubility Information | Soluble in water. |
| IUPAC Name | manganese(2+) hydrate sulfate |
| Formula Weight | 151.00 (Anhydrous) |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Appearance | Pink Crystalline Powder |
| Show More |
Safety and Handling
| Hazard Category | H373 |
| Precautionary Statement | P260-P314-P501c |
| DOTInformation | DOT Class: 9, Packaging Group: III |
| EINECSNumber | 232-089-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.