Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiogenin Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB396464 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396464 0.025 mg
NBP241185 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396464 Supplier Novus Biologicals Supplier No. NBP2411850.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

Angiogenin Polyclonal specifically detects Angiogenin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence.

Specifications

Antigen Angiogenin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 μg/mL, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 20 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias ALS9, angiogenin, angiogenin, ribonuclease, RNase A family, 5, EC 3.1.27, EC 3.1.27.-, epididymis luminal protein 168, HEL168, MGC22466, MGC71966, Ribonuclease 5, RNase 5, RNASE5RNASE4
Gene Symbols ANG
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide near the center of human ANG. The immunogen is located within amino acids 70 - 120 of ANG. Amino Acid Squence: NKNGNPHRENLRISKS
Molecular Weight of Antigen 16 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 283
Test Specificity Two alternatively spliced transcript variants have been observed.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.