Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiogenin Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$196.10 - $499.50
Specifications
Antigen | Angiogenin |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1 - 2 μg/mL, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 20 μg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Description
Angiogenin Polyclonal specifically detects Angiogenin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence.Specifications
Angiogenin | |
Western Blot 1 - 2 μg/mL, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
Polyclonal | |
Rabbit | |
Angiogenesis, Cancer | |
PBS with 0.02% Sodium Azide | |
283 | |
Antibody was raised against a 15 amino acid synthetic peptide near the center of human ANG. The immunogen is located within amino acids 70 - 120 of ANG. Amino Acid Squence: NKNGNPHRENLRISKS | |
Primary | |
Two alternatively spliced transcript variants have been observed. |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
ALS9, angiogenin, angiogenin, ribonuclease, RNase A family, 5, EC 3.1.27, EC 3.1.27.-, epididymis luminal protein 168, HEL168, MGC22466, MGC71966, Ribonuclease 5, RNase 5, RNASE5RNASE4 | |
ANG | |
IgG | |
Affinity Purified | |
16 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title