Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-1 Antibody (09), Novus Biologicals™
SDP

Catalog No. NB287429 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287429 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287429 Supplier Novus Biologicals Supplier No. NBP323500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Angiopoietin-1 Monoclonal antibody specifically detects Angiopoietin-1 in Human samples. It is validated for ELISA

Specifications

Antigen Angiopoietin-1
Applications ELISA
Classification Monoclonal
Clone 9
Conjugate Unconjugated
Dilution ELISA 1:1000-1:2000
Formulation 0.2 um filtered solution in PBS
Gene Alias AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, KIAA0003
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Angiopoietin-1 (Accession#: NP_001137.2; Met1-Leu261 and Arg277-Phe498).
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 284
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.