Learn More
Angiopoietin-1 Antibody (MM0019-26B10) - Azide and BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Angiopoietin-1 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | MM0019-26B10 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:400, Immunohistochemistry-Paraffin 1:50-1:400, Immunohistochemistry-Frozen |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. 20 ug size is provided in liquid form, PBS with No Preservative |
| Gene Accession No. | Q15389 |
| Gene Alias | AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, KIAA0003 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.