Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-1 Antibody (MM0019-26B10) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 1 publication

$231.00

Specifications

Antigen Angiopoietin-1
Clone MM0019-26B10
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB414201
SDP
View Documents
Novus Biologicals
NB1108546420UG
20 μg
Each for $231.00
Only null left
Add to Cart
 
NB11085464
SDP
View Documents
Novus Biologicals
NB11085464
0.1 mg N/A N/A N/A
Description

Description

Angiopoietin-1 Monoclonal specifically detects Angiopoietin-1 in Human, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Angiopoietin-1
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
Mouse
Angiogenesis, Cancer
Q15389
284
Primary
Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
MM0019-26B10
Monoclonal
Purified
RUO
Human, Chicken
AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, KIAA0003
ANGPT1
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.