Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-2 Antibody (MM0020-1F29) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018035 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018035 0.025 mg
NB11085467 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018035 Supplier Novus Biologicals Supplier No. NB110854670.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 4 publications

Angiopoietin-2 Monoclonal specifically detects Angiopoietin-2 in Human, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Angiopoietin-2
Applications Immunofluorescence, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone MM0020-1F29
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:25, Immunohistochemistry-Paraffin 1:100-200, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. O15123
Gene Alias AGPT2, ANG2, ANG-2, angiopoietin 2, angiopoietin-2, angiopoietin-2a, angiopoietin-2B, angiopoitin 2, Tie2-ligand
Gene Symbols ANGPT2
Host Species Mouse
Immunogen purified human recombinant Ang2
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 285
Test Specificity This recognizes human Ang2 without cross reactivity with human Ang1.
Reconstitution Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Human, Chicken
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.