Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Angiopoietin-2 Antibody (MM0020-1F29) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 4 publications
$206.50
Specifications
Antigen | Angiopoietin-2 |
---|---|
Clone | MM0020-1F29 |
Concentration | LYOPH |
Applications | Immunofluorescence, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018035
![]() |
Novus Biologicals
NB110854670.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB11085467
![]() |
Novus Biologicals
NB11085467 |
0.1 mg | N/A | N/A | N/A | ||||
Description
Angiopoietin-2 Monoclonal specifically detects Angiopoietin-2 in Human, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Angiopoietin-2 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Human, Chicken | |
AGPT2, ANG2, ANG-2, angiopoietin 2, angiopoietin-2, angiopoietin-2a, angiopoietin-2B, angiopoitin 2, Tie2-ligand | |
ANGPT2 | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
MM0020-1F29 | |
Immunofluorescence, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Mouse | |
Angiogenesis, Cancer | |
O15123 | |
285 | |
IgG2 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title