Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-4 Antibody (MM0090-11M11), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP212037C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212037C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212037C Supplier Novus Biologicals Supplier No. NBP212037C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Angiopoietin-4 Monoclonal specifically detects Angiopoietin-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Angiopoietin-4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0090-11M11
Conjugate DyLight 650
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AGP4, ANG3, ANG-3, ANG-4, ANG4angiopoietin-3, angiopoietin 4, Angiopoietin-3, angiopoietin-4, dJ824F16.2 (angiopoietin 4), MGC138181, MGC138183
Gene Symbols ANGPT4
Host Species Mouse
Immunogen Human recombinant Angiopoietin-4
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 51378
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.