Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-4 Antibody (RM0073-2H34), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP121700V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP121700V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP121700V Supplier Novus Biologicals Supplier No. NBP121700V
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Angiopoietin-4 Monoclonal specifically detects Angiopoietin-4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Angiopoietin-4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RM0073-2H34
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AGP4, ANG3, ANG-3, ANG-4, ANG4angiopoietin-3, angiopoietin 4, Angiopoietin-3, angiopoietin-4, dJ824F16.2 (angiopoietin 4), MGC138181, MGC138183
Gene Symbols ANGPT4
Host Species Rat
Immunogen Mouse recombinant Angiopoietin-3
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 51378
Test Specificity This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant Angiopoietin-3.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.