Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-like Protein 1/ANGPTL1 Antibody (OTI1B2), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP271414V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271414V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271414V Supplier Novus Biologicals Supplier No. NBP271414V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Angiopoietin-like Protein 1/ANGPTL1 Monoclonal specifically detects Angiopoietin-like Protein 1/ANGPTL1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Angiopoietin-like Protein 1/ANGPTL1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1B2
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ANG-3, ANG3angioarrestin, angiopoietin Y1, angiopoietin-3, angiopoietin-like 1, Angiopoietin-like protein 1, angiopoietin-related protein 1, ANGPT3, AngY, ARP1angiopoietin 3, dJ595C2.2, dJ595C2.2 (angiopoietin Y1), KIAA0351, UNQ162
Gene Symbols ANGPTL1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 24-491 of human Angiopoietin-like Protein 1/ANGPTL1 (NP_004664) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9068.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.