Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Angiopoietin-like Protein 3/ANGPTL3 Antibody (23D8), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB308663 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308663 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308663 Supplier Novus Biologicals Supplier No. NBP260244MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Angiopoietin-like Protein 3/ANGPTL3 Monoclonal antibody specifically detects Angiopoietin-like Protein 3/ANGPTL3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Angiopoietin-like Protein 3/ANGPTL3
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 23D8
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ANG-5, angiopoietin 5, Angiopoietin-5, angiopoietin-like 3, Angiopoietin-like protein 3, angiopoietin-related protein 3, ANGPT5angiopoietin-5, FHBL2
Host Species Mouse
Immunogen Recombinant human Angiopoietin-like Protein 3/ANGPTL3 (243-460aa) purified from E. coli.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27329
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.